missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper(I) oxide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197420025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1800.0°C | |
1232.0°C | |
1317-39-1 | |
Powder | |
96% min. (Complexometry) | |
Cu2O | |
15, 2654 | |
Solubility in water: insoluble. Other solubilities: soluble in ammonia or min.acids | |
143.09 | |
Glass bottle |
Copper(I) oxide | |
97% | |
Brown-Red | |
2.5 kg | |
Cu2O | |
MFCD00010974 | |
Cuprous oxide | |
143.09 | |
97% |
Chemical Identifiers
1317-39-1 | |
143.09 | |
Cuprous oxide |
Cu2O | |
MFCD00010974 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 215-270-7
RTECSNumber : GL8050000
TSCA : TSCA
RUO – Research Use Only