missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) iodide, 99.995%, (trace metal basis), ACROS Organics™

Manufacturer:  Acros Organics AC201501000

 View more versions of this product

Catalog No. AC201501000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

C-X bond formation reagent, for ultra trace analysis
Specifications

Specifications

Copper(I) iodide
7681-65-4
CuI
MFCD00010978
15, 2652
Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol
190.45
31.0 to 34.0% (Cu)
5.62
50ppm max.
Brown or Gray to White
100g
99.995%
99.995%
CuI
01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88
copper i iodide 250g
190.45
Powder
Trace Metal Basis
Glass bottle
1290°C
605°C

Chemical Identifiers

7681-65-4
190.45
copper i iodide 250g
CuI
MFCD00010978
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents