Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(I) hexafluoro-2,4-pentanedionate 1,5-cyclooctadiene complex

Used in MOCVD as a source of Cu2O

$179.31 - $179.31

Chemical Identifiers

Chemical Name or Material Copper(I) hexafluoro-2,4-pentanedionate 1,5-cyclooctadiene complex
CAS 86233-74-1
Formula Weight 378.77
MDL Number MFCD00156517
Synonym Copper(I) hexafluoroacetylacetonate complex with 1,5-cyclooctadiene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4191604

  SDS
alfa aesar™
4191604
2g Each for $179.31
Description & Specifications

Specifications

Formula Weight 378.77
CAS 86233-74-1
Health Hazard 1 Warning
Chemical Name or Material Copper(I) hexafluoro-2,4-pentanedionate 1,5-cyclooctadiene complex
Color Yellow-green
MDL Number MFCD00156517
Boiling Point 60°C (0.1mmHg) (subl.)
Physical Form Powder/Aggregate
Melting Point 93° to 100°C
Synonym Copper(I) hexafluoroacetylacetonate complex with 1,5-cyclooctadiene
Solubility Information Soluble in THF, hexane
Quantity 2g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.