missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) cyanide, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 202082500

 View more versions of this product

Catalog No. AC202082500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Copper(I) cyanide
544-92-3
Powder
98.5% min. (ex Cu)
CuCN
05,166; 06,146; 09,124; 16,97
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
DOBRDRYODQBAMW-UHFFFAOYSA-N
copper(1+);cyanide
11009
99%
Plastic bottle
2.9200g/mL
474.0°C
Beige-White to Green
250 g
CCuN
MFCD00010975
15, 2651
Solubility in water: practically insoluble in water. Other solubilities: soluble in acids and alkalies, practically insoluble in alcohol
[C-]#N.[Cu+]
89.56
89.56
Extra Pure
2.92

Chemical Identifiers

544-92-3
89.56
DOBRDRYODQBAMW-UHFFFAOYSA-N
11009
[C-]#N.[Cu+]
CCuN
MFCD00010975
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
copper(1+);cyanide
Safety and Handling

Safety and Handling

GHS H Statement
Fatal in contact with skin.
Fatal if inhaled.
Fatal if swallowed.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Rinse mouth.

GHS Signal Word: Danger

EINECSNumber : 208-883-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only