missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(I) chloride, 97+%, extra pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 270525000

 View more versions of this product

Catalog No. AC270525000



Description

Description

Specifications

Specifications

Copper(I) chloride
97.0 to 103% (Complexometry)
CuCl
15,265
5ppm max.
OXBLHERUFWYNTN-UHFFFAOYSA-M
chlorocopper
62652
99
97+%
4.1400g/mL
100ppm max.
Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh, forming complexes, practically insoluble in alcohol and acetone
0.5% max.
1490.0°C
430.0°C
7758-89-6
ClCu
01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97
chlorid medny czech, copper 1+ chloride, copper chloride cucl, copper i chloride, copper monochloride, cucl, cuprous chloride, dicopper dichloride
Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh,forming complexes,practically insoluble in alcohol and acetone
Cl[Cu]
99
CHEBI:53472
Crystalline Powder
Extra Pure
0.05% max. (in acid)
Glass bottle
4.14
500ppm max.
Gray to Green
500g

Chemical Identifiers

7758-89-6
99
chlorid medny czech, copper 1+ chloride, copper chloride cucl, copper i chloride, copper monochloride, cucl, cuprous chloride, dicopper dichloride
CHEBI:53472
Cl[Cu]
ClCu
OXBLHERUFWYNTN-UHFFFAOYSA-M
62652
chlorocopper
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut

GHS Signal Word: Danger

EINECSNumber : 231-842-9

SDS Documents