Please login to your online account to display your discounted pricing

Copper(I) chloride, 97%, Alfa Aesar™

$65.73 - $65.73

Chemical Identifiers

Chemical Name or Material Copper(I) chloride
CAS 7758-89-6
Molecular Formula CuCl
Formula Weight 98.99
MDL Number MFCD00010971
UN Number UN2802
Merck Index 14,2660
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1187136

  SDS
alfa aesar
1187136
Each for $65.73
Description & Specifications

Specifications

Formula Weight 98.99
Quantity 500g
Chemical Name or Material Copper(I) chloride
TSCA TSCA
Physical Form Powder
Refractive Index 1.93
Assay 97%
Sensitivity Air and Moisture Sensitive
Molecular Formula CuCl
MDL Number MFCD00010971
CAS 7758-89-6
UN Number UN2802
Merck Index 14,2660
Boiling Point 1490°C
Color Gray to Green
Melting Point 430°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302;
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.14
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: COPPER CHLORIDE
RTECS Number GL6990000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a;
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302;
Harmful if swallowed.