Please login to your online account to display your discounted pricing

Copper(I) bromide-dimethyl sulfide complex, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC213181000

 View more versions of this product

Catalog No. AC213181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Color Off-white to beige, gray or green
Physical Form Crystalline Powder and/or Chunks
Formula Weight 205.58
Quantity 100g
EINECS Number 259-292-5
Chemical Name or Material Copper(I) bromide-dimethyl sulfide complex
MDL Number MFCD00043295
Health Hazard 1 MOISTURE SENSITIVE
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula (CH3)2S·CuBr
CAS 54678-23-8
Name Note 99%
Molecular Formula C2H6BrCuS
Packaging Glass bottle
Health Hazard 2 AIR SENSITIVE

Specifications

Chemical Name or Material Copper(I) bromide-dimethyl sulfide complex
Name Note 99%
CAS 54678-23-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C2H6BrCuS
Linear Formula (CH3)2S·CuBr
Formula Weight 205.58
MDL Number MFCD00043295
Accessories

MOISTURE SENSITIVE

AIR SENSITIVE

SDS & Safety Documents