missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper(I) bromide-dimethyl sulfide complex, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 213180100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Copper(I) bromide-dimethyl sulfide complex | |
54678-23-8 | |
98.5% min. (Iodometry) | |
C2H6BrCuS | |
MFCD00043295 | |
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i | |
CSC.[Cu]Br | |
205.58 | |
205.58 |
99% | |
Authentic | |
Glass bottle | |
(CH3)2S·CuBr | |
10g | |
PMHQVHHXPFUNSP-UHFFFAOYSA-M | |
bromocopper;methylsulfanylmethane | |
2724234 | |
99% |
Safety and Handling
AIR SENSITIVE
MOISTURE SENSITIVE
EINECSNumber : 259-292-5
RUO â Research Use Only