missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper(I) bromide-dimethyl sulfide complex

Manufacturer:  Alfa Aesar™ 1936818

 View more versions of this product

Catalog No. AA1936818



Specifications

Specifications

Copper(I) bromide-dimethyl sulfide complex
C2H6BrCuS
UN3335
PMHQVHHXPFUNSP-UHFFFAOYSA-M
bromocopper;methylsulfanylmethane
2724234
Crystalline
Brown or Green
50g
54678-23-8
MFCD00043295
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i, bromo dimethylsulfide copper i
CSC.[Cu]Br
205.58
205.58
Air & Moisture Sensitive
124°C to 129°C (decomposition)

Chemical Identifiers

54678-23-8
205.58
PMHQVHHXPFUNSP-UHFFFAOYSA-M
2724234
CSC.[Cu]Br
C2H6BrCuS
MFCD00043295
copper i bromide-dimethyl sulfide, bromo thiobis methane copper, copper i bromide dimethyl sulfide complex, bromocopper; methylsulfanylmethane, cubr.me2s, ksc493e2t, copper i bromide dimethylsulfide, copper i bromide-dimethylsulfide, bromo dimethylsulfide copper i, bromo dimethylsulfide copper i
bromocopper;methylsulfanylmethane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.

SDS Documents