Please login to your online account to display your discounted pricing

Copper(I) acetate, 90%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC378810050

 View more versions of this product

Catalog No. AC378810050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥88% (Complexometry)
Formula Weight 122.59
Physical Form Powder
Quantity 5g
Color White to slightly yellow
Merck Index 15, 2648
Linear Formula CH3C(O)OCu
Assay Percent Range 90%
CAS 598-54-9
Chemical Name or Material Copper(I) acetate
MDL Number MFCD00058908
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Synonym Acetic acid, copper(I) salt
Infrared Spectrum Authentic
Molecular Formula C2H3CuO2

Specifications

Chemical Name or Material Copper(I) acetate
Assay ≥88% (Complexometry)
Assay Percent Range 90%
Molecular Formula C2H3CuO2
Linear Formula CH3C(O)OCu
Formula Weight 122.59
MDL Number MFCD00058908
Synonym Acetic acid, copper(I) salt
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents