Please login to your online account to display your discounted pricing

Copper, Granular, 20 to 30 Mesh, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$354.95 - $354.95

Chemical Identifiers

Chemical Name or Material Copper
CAS 7440-50-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Cu
Formula Weight 63.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002965

 
avantor performance
172001
Poly Bottle 500g Each for $354.95
Description & Specifications

Specifications

Formula Weight 63.55
Physical Form Granular
Molecular Formula Cu
CAS 7440-50-8
Chemical Name or Material Copper
Purity Grade BAKER ANALYZED™ Reagent
Mesh Size 20 to 30 Mesh
Packaging Poly Bottle
Specific Gravity 1L = 8.94kg
Quantity 500g