Learn More
Copper germanium selenide, 99.99% (metals basis), Thermo Scientific Chemicals
$383.49 - $383.49
Chemical Identifiers
CAS | 54723-90-9 |
---|---|
Molecular Formula | CuGeSe |
Molecular Weight (g/mol) | 215.15 |
MDL Number | MFCD12546179 |
InChI Key | LQTVKWORTHHMFT-UHFFFAOYSA-N |
Synonym | copper germanium selenide |
PubChem CID | 121233948 |
IUPAC Name | selanylidenegermylidenecopper |
SMILES | [Cu]=[Ge]=[Se] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4560509
|
Thermo Scientific Chemicals
04560509 |
10 g |
Each for $383.49
|
|
|||||
Description
The most notable physical characteristics of germania (GeO2) are its high index of refraction and its low optical dispersion. These make it especially useful for wide-angle camera lenses, microscopy, and for the core part of optical fibers. Germanium-on-insulator substrates are seen as a potential replacement for silicon on miniaturized chips.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54723-90-9 | |
215.15 | |
LQTVKWORTHHMFT-UHFFFAOYSA-N | |
121233948 | |
[Cu]=[Ge]=[Se] |
CuGeSe | |
MFCD12546179 | |
copper germanium selenide | |
selanylidenegermylidenecopper |
Specifications
54723-90-9 | |
10 g | |
MFCD12546179 | |
copper germanium selenide | |
LQTVKWORTHHMFT-UHFFFAOYSA-N | |
selanylidenegermylidenecopper | |
121233948 | |
99.99% | |
(metals basis) |
Pieces | |
CuGeSe | |
UN3283 | |
Insoluble in water. | |
[Cu]=[Ge]=[Se] | |
215.15 | |
215.12 | |
Copper germanium selenide |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only