missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper germanium selenide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4560518

 View more versions of this product

Catalog No. AA4560518



Specifications

Specifications

Copper germanium selenide
(metals basis)
MFCD12546179
copper germanium selenide
[Cu]=[Ge]=[Se]
215.147
215.12
99.99%
50g
54723-90-9
CuGeSe
UN3283
LQTVKWORTHHMFT-UHFFFAOYSA-N
selanylidenegermylidenecopper
121233948
Pieces
3 to 6mm

Chemical Identifiers

54723-90-9
215.147
LQTVKWORTHHMFT-UHFFFAOYSA-N
121233948
[Cu]=[Ge]=[Se]
CuGeSe
MFCD12546179
copper germanium selenide
selanylidenegermylidenecopper
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.

SDS Documents