missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper germanide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4560418

 View more versions of this product

Catalog No. AA4560418

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Copper germanide
CuGe
136.16
99.99%
50g
(metals basis)
MFCD12546178
Pieces
3 to 6mm

Chemical Identifiers

CuGe
MFCD12546178
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents