missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper gauze, 100 mesh woven from 0.11mm (0.0045in) dia wire, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045186HB
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper | |
Copper colored guaze | |
Cu | |
300 x 300 mm | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
23978 | |
100 Mesh | |
0 hPa |
7440-50-8 | |
8.94g/cm3 | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
63.55 | |
CHEBI:30052 | |
Odorless | |
Gauze |
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Safety and Handling
EINECSNumber : 231-159-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only