missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Gauze, 60 mesh woven from 0.19mm (0.0075 in.) dia wire, , Alfa Aesar™

Manufacturer:  Alfa Aesar 45142HB

 View more versions of this product

Catalog No. AA45142HB



Specifications

Specifications

Copper
7440-50-8
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
Odorless
copper
23978
Gauze
300 x 300mm
Copper colored guaze
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
[Cu]
63.546
CHEBI:30052
Undesignated

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
SDS Documents