missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper gauze, 30 mesh woven from 0.15mm (0.006 in.) dia. wire

Manufacturer:  Alfa Aesar™ 46834HB

 View more versions of this product

Catalog No. AA46834HB



Specifications

Specifications

Copper gauze
30 mesh woven from 0.15mm (0.006 in.) dia. wire
Cu
30 Mesh
RYGMFSIKBFXOCR-UHFFFAOYSA-N
[Cu]
63.546
CHEBI:30052
300 x 300mm
7440-50-8
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
Odorless
copper
23978
Wire Cloth

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents