missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper gauze, 140 mesh woven from 0.056mm (0.0022 in.) dia. wire

Manufacturer:  Alfa Aesar™ 46603FL

 View more versions of this product

Catalog No. AA46603FL



Specifications

Specifications

Copper gauze
140 mesh woven from 0.056mm (0.0022 in.) dia. wire
Cu
140 Mesh
Odorless
copper
23978
Wire Cloth
75 x 75mm
7440-50-8
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
[Cu]
63.546
CHEBI:30052

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
CHEBI:30052
[Cu]
Cu
MFCD00010965
23978
copper
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents