missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper gauze, 10 mesh woven from 0.15mm (0.006 in.) dia. wire

Manufacturer:  Alfa Aesar™ 46515HB

 View more versions of this product

Catalog No. AA46515HB



Specifications

Specifications

Copper gauze
10 mesh woven from 0.15mm (0.006 in.) dia. wire
Cu
10 Mesh
Odorless
copper
23978
Wire Cloth
300 x 300mm
7440-50-8
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
[Cu]
63.546
CHEBI:30052

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
CHEBI:30052
[Cu]
Cu
MFCD00010965
23978
copper
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents