missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper foil, Oxygen-Free High Conductivity (OFHC), alloy 101, 1.6mm (0.064in) thick, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045168GT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper | |
7440-50-8 | |
8.94g/cm3 | |
MFCD00010965 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
[Cu] | |
63.55 | |
CHEBI:48107 | |
Foil |
Oxygen-Free High Conductivity (OFHC), alloy 101, 1.6mm (0.064 in.) thick | |
Copper colored foil | |
Cu | |
150 x 150 mm | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
nitric acid | |
944 | |
Odorless |
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
944 | |
[Cu] |
Cu | |
MFCD00010965 | |
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
CHEBI:48107 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only