missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper foil, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 1.1g/25x200mm

Manufacturer:  Alfa Aesar 10950GZ

 View more versions of this product

Catalog No. AA10950GZ



Specifications

Specifications

Copper
7440-50-8
8.94g/cm3
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Odorless
23978
Foil, 0.025mm (0.001 in.) thick
Undesignated
25 x 1000mm
Copper colored foil
99.999% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
0 hPa

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

Documents