missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper foil, 99.99% (metals basis), Alfa Aesar™

Form: approx. 2.2g/50x50mm

Manufacturer:  Thermo Fisher Scientific 10954FI

 View more versions of this product

Catalog No. AA10954FI



Specifications

Specifications

Copper
7440-50-8
99.9999% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
0 hPa
50 x 50mm
Copper colored foil
Cu
anode
[Cu]
Odorless
23978
Foil, 1.0mm (0.04 in.) thick
Undesignated

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

Documents