missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper foil, 1.0mm (0.04 in.) thick, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 11378HL

 View more versions of this product

Catalog No. AA11378HL



Specifications

Specifications

Copper foil
100 x 250mm
7440-50-8
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Odorless
23978
Foil
Puratronic™
≈11.2g/25x50mm
1.0mm (0.04 in.) thick
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
99.999%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents