missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated, 99.8% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 46986RH

 View more versions of this product

Catalog No. AA46986RH



Specifications

Specifications

Copper foil
7440-50-8
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
30 x 1000cm
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Shrink-wrapped
23978
99.8%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : Yes

SDS Documents