missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper flake, -325 mesh, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4507330

 View more versions of this product

Catalog No. AA4507330



Specifications

Specifications

Copper
7440-50-8
99% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
[Cu]
63.546
CHEBI:30052
Flakes
Undesignated
250g
Copper colored flakes
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
Odorless
copper
23978
63.55
0 hPa

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

H228-H319-H335-H411-H401: Flammable solid. Causes serious eye irritation. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-159-6

SDS Documents