missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper discs, 9.0+/-0.127mm (0.354+/-0.005 in.) dia. x 0.8+/-0.051mm (0.03+/-0.002 in.) thick, 99.9995% (metals basis)

Manufacturer:  Alfa Aesar™ 1095109

 View more versions of this product

Catalog No. AA1095109



Specifications

Specifications

Copper discs
7440-50-8
Cu
anode
[Cu]
63.546
CHEBI:30052
Odorless
≈0.5g/disc
9.0±0.127mm (0.354±0.005 in.) dia. x 0.8±0.051mm (0.03±0.002 in.) thick
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
99.9995%
10g

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents