Please login to your online account to display your discounted pricing

Copper chromite, catalyst, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Copper chromite
Name Note Catalyst
CAS 12018-10-9
Assay 36% min. (Cu) (on dry substance), 26% min. (Cr) (on dry substance)
Molecular Formula Cr2Cu2O5
Linear Formula 2CuO·Cr2O3
Formula Weight 311.07g/mol
MDL Number MFCD00064661
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC197821000

  SDS
thermo fisher scientific
197821000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Loss on Drying 0.5% max.
Packaging Plastic bottle
Color Brown to black
pH 5 to 7 (10% susp.)
Quantity 100g
Chemical Name or Material Copper chromite
Name Note Catalyst
CAS 12018-10-9
Physical Form fine brown to black powder
Assay 36% min. (Cu) (on dry substance), 26% min. (Cr) (on dry substance)
Molecular Formula Cr2Cu2O5
Linear Formula 2CuO·Cr2O3
Formula Weight 311.07g/mol
MDL Number MFCD00064661
Fieser 01,156; 02,82
Merck Index 15, 2625
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.