missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 47162MD

 View more versions of this product

Catalog No. AA47162MD



Specifications

Specifications

Copper bis(trifluoromethylsulfonyl)imide
250mg
Hygroscopic
[(CF3SO2)2N]Cu
UN3261
MSFHCZUIBARZOS-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;copper(1+)
134159300
Solid
291300-50-0
Blue
C2CuF6NO4S2
MFCD23380170
copper trifluoromethanesulfonimide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cu+]
343.681
343.68

Chemical Identifiers

291300-50-0
343.681
MSFHCZUIBARZOS-UHFFFAOYSA-N
134159300
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cu+]
C2CuF6NO4S2
MFCD23380170
copper trifluoromethanesulfonimide
bis(trifluoromethylsulfonyl)azanide;copper(1+)
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents