Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper Beryllium foil, alloy C17200, 0.15mm (0.006 in.) thick

Chemical Identifiers

Chemical Name or Material Copper Beryllium foil
Name Note Alloy C17200, 0.15mm (0.006 in.) thick
MDL Number MFCD01320550
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41912GT

  SDS
alfa aesar™
41912GT
150x150mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Copper Beryllium foil
TSCA TSCA
Name Note Alloy C17200, 0.15mm (0.006 in.) thick
Physical Form Foil
MDL Number MFCD01320550
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H373-H332-H317
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Be; 98.1:1.9 wt%
Weight ≈37.7g/150x200mm
Quantity 150x150mm
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H373-H332-H317
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
May cause an allergic skin reaction.