missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper, ACS Reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC448055000

 View more versions of this product

Catalog No. AC448055000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Copper
7440-50-8
Cu
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
Glass bottle
CHEBI:30052
ACS Reagent
2595.0°C
500g
Cu
anode, blister, bronze, cathode, cobre, cuivre, cuprum, kupfer, powder, precipitates
[Cu]
63.546
23978
63.54
1083.0°C

Chemical Identifiers

7440-50-8
63.546
anode, blister, bronze, cathode, cobre, cuivre, cuprum, kupfer, powder, precipitates
CHEBI:30052
[Cu]
Cu
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
SDS Documents