Copan Diagnostics T-Shaped Spreaders
T-shaped spreaders
Supplier:  Copan Diagnostics COPTS1
 View more versions of this product
Catalog No. 23-600-894
Description
Description
T-shaped spreaders
T-Shaped Plastic Spreaders
  • Designed for spreading and dispersing liquids across the surface of agar culture plates
  • Used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples
  • Smooth, rounded surfaces prevent any cutting or gouging of agar during inoculation of culture plates
  • T-Shape design means that even pressure is applied across the entire length of the T-bar in contact with the agar
  • Available individually sterile wrapped or in packs of five or ten pieces in a peel pack
Specifications
Specifications
Sterile
1/peel pack; 500 spreaders/box
Plastic
Designed for spreading and dispersing liquids across the surface of agar culture plates, used for performing bacterial CFU plate counts on water, milk, urine and other liquid samples
T-Shape
1/peel pk.; 500 Spreaders/Box
Culture Plates