Please login to your online account to display your discounted pricing
Copan Diagnostics CRYOBANK™Bead System
Easy-to-use system for freezing bacterial cultures for later retrieval
$356.25 - $356.25
Specifications
For Use With (Application) For freezing bacterial cultures for later retrieval
Quantity 64/Pk.
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
14-906-51
copan diagnostics
CRYOM
Assorted Colors Pack of 64 Each for $356.25
Description & Specifications
Specifications
For Use With (Application) For freezing bacterial cultures for later retrieval
Quantity 64/Pk.
  • Cryogenic medium consists of a phosphate buffered saline supplemented with sucrose, glycerol, and peptone
  • Can be stored for long periods at -60° to -80°C
  • Easy to store in unique storage boxes designed to fit freezer racks
  • Comes with 64 vials
  • Easy to locate
  • Convenient, versatile and reliable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.