missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copan Diagnostics Calibrated Disposable Inoculating Loops and Needles in Zip Lock Pack

Calibrated 1 and 10μL plastic disposable inoculating loops and needles

Manufacturer:  Copan Diagnostics COPNED

 View more versions of this product

Catalog No. OXCOPNED



Description

Description

Calibrated 1 and 10μL plastic disposable inoculating loops and needles

Zip-lock closure reseals the pack between use to avoid contamination. Re-sealable bag allows the user to open and close the loop pack to remove individual loops.

  • Disposable, sterile plastic loops and needles manufactured from soft-flexible or hard-rigid plastic
  • Plastic injection molding technology ensures product consistency and eliminates variations in loop thickness and internal diameter
  • Ultrasmooth surfaces allow easy streaking without cutting or gouging agar surface
  • Calibrated using the Evans Blue Dye Method recommended by the American Society for Microbiology
  • Color coded for easy product identification
  • Packed in tamper-proof bags to guarantee sterility
  • Certificate of calibration included in each case

Specifications

Specifications

Sterile
Violet
0.14cm
Evans blue dye certificate of calibration
20cm
Tamperproof Ziplock resealable bag
Inoculating Needle
0.057 in.
Yes
7.84 in.
Plastic
Yes
Documents