Please login to your online account to display your discounted pricing

CoorsTek™ Spot Plates

For observing color reactions

$91.07 - $413.00

Specifications

Color White
For Use With (Application) For observing color reactions
View More Specs
Products
Catalog Number Mfr. No. Length (English) No. of Wells Width (English) Price Quantity    

13-745-5

 
coorstek™
CRS60427
Each for $111.33
Case of 12 Each for $1,335.96

13-745-8

 
coorstek™
CRS60430
6.125 in. 12 4.6 in. Each for $207.00
Case of 8 Each for $1,656.00

13-745-9

 
coorstek™
CRS60432
7 in. 30 5.5 in. Each for $413.00
Case of 6 Each for $2,478.00

13-745-12A

 
coorstek™
CRS60425
3.6 in. 3 1.2 in. Each for $91.07
Case of 24 Each for $2,185.68

13-745-12B

 
coorstek™
CRS60429
4.6 in. 12 3.5 in. Each for $113.27
Case of 12 Each for $1,359.24
Description & Specifications

Specifications

Color White
For Use With (Application) For observing color reactions

Glazed except for bottom surfaces.