CoorsTek™ Porcelain Gooch Filtering Crucibles
Glazed inside and out except for outside bottom surface
$50.97 - $245.67
Specifications
Product Type Specs Gooch Filtering Crucible
Finish Glazed
For Use With (Application) For total suspended solids determination
Material Porcelain
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
36-104-0137
View Documents Promotion Details
CoorsTek™
CRS60151
40 mL
Each for $50.97
08-195B
Accessories View Documents Promotion Details
CoorsTek™
CRS66146
13mL
Each for $65.33
Case of 24 Each for $1,567.92
08-215-1
Accessories View Documents Promotion Details
CoorsTek™
CRS66153
130mL
Each for $245.67
Case of 6 Each for $1,474.02
Description
Description
  • Perforated bottoms
  • Perforations, 0.013 in. dia. (0.7mm) in 13 to 40mL size; 0.046 in. dia. (1mm) in 130mL size
  • Covers sold separately
Specifications
Specifications
Gooch Filtering Crucible
Glazed
For total suspended solids determination
Porcelain