CoorsTek™ Porcelain Desiccator Plates for Crucibles
With 30mm diameter holes for crucibles
$152.96 - $249.17
Specifications
Material Porcelain
Holds 3
Includes Three 16mm feet
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Outer Diameter (Metric) Outer Product Type Price Quantity  
Catalog Number Mfr. No. Diameter (English) Outer Diameter (Metric) Outer Product Type Price Quantity  
08-636F CoorsTek™
CRS60449
9.05 in. 23cm Desiccator Plate w/Feet Each for $249.17
Case of 3 Each for $747.51
08-636D CoorsTek™
CRS60447
5.74 in. 14.6cm Desiccator Plate w/Feet Each for $164.09
Case of 6 Each for $984.54
08-641D CoorsTek™
CRS60451
9.05 in. 23cm Desiccator Plate w/o Feet Each for $223.98
Case of 3 Each for $671.94
08-636A CoorsTek™
CRS60444
3.74 in. 9.5cm Desiccator Plate w/Feet Each for $241.41
Case of 8 Each for $1,931.28
08-636C CoorsTek™
CRS60446
5.59 in. 14.2cm Desiccator Plate w/Feet Each for $152.96
Case of 6 Each for $917.76
08-636B CoorsTek™
CRS60445
4.52 in. 11.5cm Desiccator Plate w/Feet Each for $153.00
Case of 8 Each for $1,224.00
08-636E CoorsTek™
CRS60448
7.48 in. 19cm Desiccator Plate w/Feet Each for $199.39
Case of 4 Each for $797.56
Specifications
Description
  • With or without three 16mm feet to support plate
  • Glazed on top
Description & Specifications
Specifications
Porcelain
3
Three 16mm feet