CoorsTek™ Porcelain Coal Moisture Testing Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
Manufacturer:  CoorsTek™ CRS60054
 View more versions of this product
Catalog No. 08-103D
Description
Description
  • Porcelain with rounded bottom
  • Glazed inside and out except on rim
  • 20mL size recommended for ASTM™ D 3176
  • Capsule form; without covers
Specifications
Specifications
35mL
Porcelain
SDS
Documents
Product Certifications
Accessories