CoorsTek™ Porcelain Buchner Funnel with Fixed Perforated Plate for Vacuum
One-piece table-type style
$2,168.03 - $2,956.00
Specifications
Autoclavable Yes
Material Porcelain
Product Type Buchner Funnel
For Use With (Application) Vacuum filtration
Includes Fixed perforated plate, side outlet for vacuum filtration
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (English) Diameter (English) Plate Quantity Height (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Plate Quantity Height (English) Price Quantity  
10-354G CoorsTek™
CRS60281
25.3 oz. 4.9 in. 2 Pack 4.29 in. Each for $2,168.03
Case of 2 Each for $4,336.06
10-354H CoorsTek™
CRS60282
64.2 oz. 7.3 in. Each 4.8 in. Each for $2,956.00
Specifications
Description
  • Side outlet for vacuum filtration
  • Glazed inside and out except for rim
Description & Specifications
Specifications
Yes
Buchner Funnel
Fixed perforated plate, side outlet for vacuum filtration
Porcelain
Vacuum filtration