Please login to your online account to display your discounted pricing
CoorsTek™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
$12.50 - $12.50
Products
Catalog Number Mfr. No. Price Quantity    
S32603CR
coorstek™
60103EMD
Each for $12.50
Description & Specifications
  • Glazed inside and out except for outside bottom surface and rim
  • Covers sold separately