CoorsTek™ Aluminum Oxide Mortars and Pestles
Extra hardness and abrasion resistance compared to agate
Manufacturer:  CoorsTek™ CRS60361
 View more versions of this product
Catalog No. 12-948C
Specifications Description & Specifications
Specifications
Used for elemental analysis, basic laboratory purposes and sample preparation and emulsifications
15mL
0.5 oz.
3.1cm
2.55 in.
1.22 in.
6.5cm
Alumina
Glazed outside
0.5 oz., 15mL
Mortar
Accessories