Please login to your online account to display your discounted pricing

Coomassie Violet R-150, MP Biomedicals


Quantity 5g

Manufacturer: mp biomedicals  0219068305

 View more versions of this product

Catalog No. ICN19068305

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5g
Chemical Name or Material Coomassie Violet-150
Name Note Protein stain in electrophoresis.
CAS 4129-84-4
Molecular Formula C41H45N3O6S2·Na

Specifications

Chemical Name or Material Coomassie Violet-150
Name Note Protein stain in electrophoresis.
CAS 4129-84-4
Molecular Formula C41H45N3O6S2·Na