Please login to your online account to display your discounted pricing

Coomassie Violet R-150, MP Biomedicals

$68.88 - $68.88

Chemical Identifiers

Chemical Name or Material Coomassie Violet-150
Name Note Protein stain in electrophoresis.
CAS 4129-84-4
Molecular Formula C41H45N3O6S2·Na
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19068325

 
mp biomedicals
19068325
25g Each for $68.88
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Coomassie Violet-150
Name Note Protein stain in electrophoresis.
Molecular Formula C41H45N3O6S2·Na
CAS 4129-84-4