Please login to your online account to display your discounted pricing
Corning™ CoolCell™ Stem Cell Cryopreservation System
Alcohol-free cell freezing system that controls the rate of freezing to −17°C/minute when placed in a −80°C freezer.
$2,563.79 - $2,563.79
Specifications
Product Line Cryopreservation system
View More Specs
Products
Catalog Number Mfr. No. Type Price Quantity    
07-210-013
corning™
C432013
CoolCell SV10 Case for $2,563.79
07-210-012
corning™
C432012
CoolCell SV2 Case for $2,563.79
Description & Specifications
Specifications
Product Line Cryopreservation system
  • Includes XT Starter Benchtop Cooler, Corning CoolRack and CoolCell Cell freezing container for 6 injectable vials
  • Color: Purple