missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ CoolCell™ LX Cell Freezing Vial Containers

Provides alcohol-free freezing at the rate of −1°C/minute when placed in a −80°C freezer.

Manufacturer:  Corning™ C432002

 View more versions of this product

Catalog No. 07-210-002


 

Specifications

Description

Container, Vial; Corning; CoolCell LX; Controlled cell freezing; Alcohol and fluid free; High cell recovery, cell viability; Reproducible cell cryopreservation; Exposed vial top; For 1.2 - 2mL vials; Green; Capacity: 12 vials; 1/Pk.

  • Alcohol and fluid-free freezing
  • High cell recovery and cell viability
  • Reproducibility
Description & Specifications

Specifications

Green
1
Cryopreservation of most cells and cell lines
12 standard 1.0 to 2.0mL cryogenic vials
11.7 x 10.11cm
Alcohol and fluid free
1
Cell Freezing Container
Polyethylene foam and thermo-conductive alloy core
Round
12.7mm
Promotions

Promotions