missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Purus™ Controlled Environments Mat: PROMO

Removes particles from foot traffic and equipment wheels.

Manufacturer:  Purus™ PFM31503101PM

 View more versions of this product

Catalog No. 19-125-937PM



Description

Description

  • Each layer is coated with an adhesive to capture soil and contain it until the layer is peeled away and discarded
  • Numbered tabs ensure one layer at a time is removed and indicates the number of remaining layers
  • Non-skid backing is applied to the bottom of the frame to provide stability on a variety of floor surfaces
  • Frame mat can be easily relocated to high-traffic areas and removed during routine floor maintenanc
  • Protect a facility from dust, dirt and contamination
  • 30- or 60-layered mats applied to reusable polystyrene frames
  • Length 61cm (24 in.)
  • Width 76.2cm (30 in.)
Specifications

Specifications

White
61 x 76.2cm (24 x 30 in.)
30 layer double white frame
Documents
Promotions

Promotions