Contoured Chemical Goggles
Superior design offers improved optics, comfort, affordability and convenience.
Manufacturer:  Fisher Science Education 05058201EMD
 View more versions of this product
Catalog No. S47629
Description
Description
  • Curved lens provides distortion-free optics with a full 160° field of vision
  • Comfortable soft vinyl frame fits over most glasses
  • Integrated side vents are designed for optimal air passage
  • Quick-draw strap can be adjusted in seconds
Compliance
SEI certified to ANSI™ Z87.1-2003 High Impact and CSA Z94.3-2007.
Specifications
Specifications
ANS Z87.1-2003 High Impact and CSA Z94.3-2007
Vinyl
Side
Anti-fog
Education
Curved for distortion-free optics
Anti-fog
Clear
Chemical-resistant Goggles
Documents