Contec™ SATWipes™ Presaturated Wipes and Dispensers
Prepare surfaces in cleanrooms. The Contec™ SATWipes™ Presaturated Wipes are offered in various options to help tackle general-purpose cleaning in ISO Class 5 to ISO Class 8 environments.
$107.22 - $178.38
Specifications
For Use With (Application) General purpose cleaning and surface preparation
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W) Item Description Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Item Description Price Quantity  
19-033-791 Contec™
SWNW0013
9 x 7 in. (23 x 18cm) Wipes with easy dispensing bucket Case of 1400 Each for $178.38
19-033-790 Contec™
SWNW0014
5.5 x 8 in. (14 x 20cm) Wipes with center-pull canister Case of 1080 Each for $107.22
Specifications
Description
  • Durable, presaturated wipes with a choice of solvent saturations on perforated rolls
  • Polyester nonwoven wipes are ideal for critical applications in ISO Class 5 environments
  • Polyester/Cellulose nonwoven wipes offer strength and low extractables for use in less critical applications in ISO Class 6 to 8 environments
  • Designed for general-purpose cleaning and surface preparation, wiping process tools and other equipment and for use prior to cleanroom entry
Order Info
Cat. No. 19-033-790: 180 wipes/canister. Cat. No. 19-033-791: 700 wipes/bucket.
Description & Specifications
Specifications
General purpose cleaning and surface preparation