Contec™ QuickConect™ Mop Head Frames
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ Quick Connect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Manufacturer:  Contec™ 2653A
 View more versions of this product
Catalog No. 19-130-5910
Description
Description
  • Offer maximum flexibility for cleaning in critical environments.
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
  • Capped end and quick-connect feature for easy attachment
Specifications
Specifications
Designed for use in pharmaceutical and medical device manufacturing facilities, or other similar critical or controlled environments.
Large mop head frame and connector for VertiKlean
36 cm
Stainless steel frame with aluminum connector
Large VertiKlean mop heads
14 in.
Yes
Mop head frame
Documents