missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Contec™ PROSAT™ Presaturated Nonwoven Wipes
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
$14.58 - $1,018.01
Specifications
For Use With (Application) | Ideal for routine cleaning and wiping instruments and equipment |
---|---|
Product Type | PROSAT™ Presaturated Wipes |
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Saturation Solution | Packaging Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Saturation Solution | Packaging Quantity | Price | Quantity | ||
19-033-773
|
Contec™
PSPP0043 |
11 x 9 in. (28 x 23 cm) | Polypropylene | 70% IPA/ 30% DI Water | 50 wipes/bag |
Case of 2000 Each for $867.16
|
|
19-150-0704
|
Contec™
PS850 |
8 x 8 in. (20.3 x 20.3 cm) | Polypropylene | 70% IPA/ 30% DI Water | 50 wipes/bag |
Pack of 50 Each for $14.58
Case of 50 Pack for $729.00 |
|
19-150-0709
|
Contec™
PSSI |
9 x 9 in. (23 x 23 cm) | Polyester | 10% IPA/ 90% DI Water | 30 wipes/bag |
Case of 50 Each for $1,002.08
|
|
19-150-0705
|
Contec™
PSC20002 |
11 x 9 in. (28 x 23 cm) | Cellulose, Polyester | 70% IPA/ 30% DI Water | 24 wipes/bag |
Case of 1200 Each for $859.57
|
|
19-150-0707
|
Contec™
PSC20006 |
11 x 9 in. (28 x 23 cm) | Cellulose, Polyester | 70% IPA/ 30% DI Water | 50 wipes/bag |
Case of 1400 Each for $760.41
|
|
19-121-001
|
Contec™
PS919 |
11 x 9 in. (28 x 23 cm) | Polypropylene | 91% IPA/ 9% DI Water | 30 wipes/bag |
Case of 1500 Each for $838.69
|
|
19-041-638
|
Contec™
PSHS1090 |
9 x 6 in. (23 x 15 cm) | Polyester Knit | 91% IPA/ 9% DI Water | 30 wipes/bag |
Pack of 30 Each for $24.81
Case of 40 Pack for $992.40 |
|
19-033-390
|
Contec™
PSC20016 |
11 x 9 in. (28 x 23 cm) | Cellulose, Polyester | 70% IPA/ 30% DI Water | 28 wipes/bag |
Case of 1400 Each for $733.36
|
|
19-047-630
|
Contec™
PSPS0008 |
9 x 9 in. (23 x 23 cm) | Polyester Knit | 70% IPA/ 30% DI Water | 150 wipes/bucket |
Case of 300 Each for $231.45
|
|
19-033-710
|
Contec™
PSCP0001 |
6 x 9 in. (15 x 23 cm) | Cellulose, Polyester | 70% IPA/ 30% DI Water | 50 ea/bg; 8 bg/master bag; 5 master bags/ case |
Case of 2000 Each for $1,018.01
|
|
Description
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
- Wipe substrate designed for the application
- Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
- Ideal for wiping articles prior to pass through
- Made of meltblown polypropylene or cellulose/polyester
- Effective and convenient for wipe-downs of process tools and other equipment
- Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
- Solvent blend meets SEMI specifications for metals and contaminants
Specifications
Ideal for routine cleaning and wiping instruments and equipment | |
PROSAT™ Presaturated Wipes |