Contec™ Polynit™ Wipes
Clean surfaces of products and process tools. The Contec™ Polynit™ Wipes are a durable option for ISO Class 5 environments.
Manufacturer:  Contec™ PN99
 View more versions of this product
Catalog No. 19-062713
Description
Description
  • For general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment
Specifications
Specifications
9 in.
ISO Class 5
9 x 9 in. (23 x 23 cm)
9 in.
Polyester
9 in.
9 in.
23 cm
White
Cleanroom
23 cm
Polynit™ Wipes
150 wipes/bag, Stacked
Non-sterile